Assay Details
| Gene Symbol: CASC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012858.4 | XP_004012907.2 | 10 - 11 | 2454 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASC3| Gene Symbol: CASC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012858.4 | XP_004012907.2 | 10 - 11 | 2454 | 64 | |
Set Membership: |
|