Assay Details
| Gene Symbol: LOC101107295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012952.3 | XP_004013001.1 | 5 - 6 | 618 | 68 | |
| XM_012186138.3 | XP_012041528.1 | 5 - 6 | 573 | 68 | ||
More Information
Set Membership: |
|

LOC101107295
Genomic Map