Assay Details
| Gene Symbol: LOC101107295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012952.3 | XP_004013001.1 | 8 - 9 | 938 | 79 | |
| XM_012186138.3 | XP_012041528.1 | 8 - 9 | 893 | 79 | ||
More Information
Set Membership: |
|

LOC101107295
Genomic Map