Assay Details
| Gene Symbol: LGALS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978255.1 | XP_027834056.1 | 7 - 8 | 1402 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LGALS4| Gene Symbol: LGALS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978255.1 | XP_027834056.1 | 7 - 8 | 1402 | 64 | |
Set Membership: |
|