Assay Details
| Gene Symbol: LOC114118847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980240.1 | XP_027836041.1 | 8 - 9 | 889 | 67 | |
| Gene Symbol: LOC101104528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956323.1 | XP_027812124.1 | 59 - 60 | 10653 | 67 | |
More Information
Set Membership: |
|

LOC114118847
Genomic Map