Assay Details
| Gene Symbol: TASOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099687.2 | XP_014955173.2 | 18 - 19 | 7730 | 93 | |
| XM_015099688.2 | XP_014955174.2 | 18 - 19 | 7711 | 93 | ||
| XM_015099691.2 | XP_014955177.2 | 17 - 18 | 7486 | 93 | ||
| XM_027976780.1 | XP_027832581.1 | 18 - 19 | 7730 | 93 | ||
| XM_027976781.1 | XP_027832582.1 | 19 - 20 | 4160 | 93 | ||
| XM_027976782.1 | XP_027832583.1 | 12 - 13 | 2853 | 93 | ||
| XM_027976783.1 | XP_027832584.1 | 17 - 18 | 7468 | 93 | ||

TASOR2
Genomic Map