Assay Details
| Gene Symbol: LOC101110178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980021.1 | XP_027835822.1 | 5 - 6 | 644 | 109 | |
| XM_027980022.1 | XP_027835823.1 | 5 - 6 | 644 | 109 | ||
| XM_027980023.1 | XP_027835824.1 | 5 - 6 | 680 | 109 | ||
| XM_027980024.1 | XP_027835825.1 | 5 - 6 | 644 | 109 | ||
More Information
Set Membership: |
|

LOC101110178
Genomic Map