Assay Details
| Gene Symbol: LOC101110178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980022.1 | XP_027835823.1 | 9 - 10 | 936 | 76 | |
| XM_027980023.1 | XP_027835824.1 | 8 - 9 | 932 | 76 | ||
More Information
Set Membership: |
|

LOC101110178
Genomic Map