Assay Details
| Gene Symbol: LOC443423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002637.4 | XP_004002686.1 | 5 - 6 | 976 | 81 | |
| GenBank mRNA | X90567.1 | - | 674 | 81 | ||
More Information
Set Membership: |
|

LOC443423
Genomic Map