Assay Details
| Gene Symbol: FILIP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002872.4 | XP_004002921.3 | 2 - 3 | 2809 | 66 | |
| XM_012188638.3 | XP_012044028.2 | 5 - 6 | 3861 | 66 | ||
| XM_027979280.1 | XP_027835081.1 | 2 - 3 | 2987 | 66 | ||
More Information
Set Membership: |
|

FILIP1L
Genomic Map