Assay Details
| Gene Symbol: AQP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004143.4 | XP_004004192.1 | 4 - 5 | 472 | 67 | |
| XM_012118370.3 | XP_011973760.1 | 4 - 5 | 488 | 67 | ||
| XM_012118379.3 | XP_011973769.1 | 3 - 4 | 423 | 67 | ||
| XM_012118386.3 | XP_011973776.1 | 3 - 4 | 439 | 67 | ||
| XM_027964298.1 | XP_027820099.1 | 2 - 3 | 430 | 67 | ||
More Information
Set Membership: |
|

AQP7
Genomic Map