Assay Details
| Gene Symbol: POLR2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004488.4 | XP_004004537.1 | 3 - 4 | 398 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POLR2D| Gene Symbol: POLR2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004488.4 | XP_004004537.1 | 3 - 4 | 398 | 83 | |
Set Membership: |
|