Assay Details
| Gene Symbol: GALNTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969138.1 | XP_027824939.1 | 2 - 3 | 385 | 63 | |
| XM_027969139.1 | XP_027824940.1 | 3 - 4 | 468 | 63 | ||
| GenBank mRNA | KX832730.1 | - | 3 - 4 | 475 | 63 | |
More Information
Set Membership: |
|

GALNTL5
Genomic Map