Assay Details
| Gene Symbol: MARCHF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008568.4 | XP_004008617.2 | 4 - 5 | 768 | 89 | |
| XM_027969748.1 | XP_027825549.1 | 4 - 5 | 719 | 89 | ||
More Information
Set Membership: |
|

MARCHF2
Genomic Map