Assay Details
| Gene Symbol: LOC114116372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973017.1 | XP_027828818.1 | 14 - 15 | 1830 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114116372| Gene Symbol: LOC114116372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973017.1 | XP_027828818.1 | 14 - 15 | 1830 | 57 | |
Set Membership: |
|