Assay Details
| Gene Symbol: G6PC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012951.4 | XP_004013000.2 | 4 - 5 | 688 | 68 | |
| XM_012186137.3 | XP_012041527.2 | 4 - 5 | 518 | 68 | ||
| GenBank mRNA | EF062861.1 | - | 3 - 4 | 328 | 68 | |
More Information
Set Membership: |
|

G6PC
Genomic Map