Assay Details
| Gene Symbol: AMPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016161.4 | XP_004016210.2 | 5 - 6 | 848 | 57 | |
| XM_012095777.3 | XP_011951167.2 | 5 - 6 | 1294 | 57 | ||
| XM_012095778.3 | XP_011951168.2 | 5 - 6 | 868 | 57 | ||
| XM_012095779.3 | XP_011951169.2 | 5 - 6 | 973 | 57 | ||
More Information
Set Membership: |
|

AMPD3
Genomic Map