Assay Details
| Gene Symbol: IL17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018887.4 | XP_004018936.1 | 2 - 3 | 227 | 74 | |
| GenBank mRNA | LN835312.1 | - | 2 - 3 | 227 | 74 | |
More Information
Set Membership: |
|

IL17A
Genomic Map