Assay Details
| Gene Symbol: LOC101106976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018899.4 | XP_004018948.3 | 5 - 6 | 485 | 98 | |
| XM_012100855.3 | XP_011956245.1 | 5 - 6 | 561 | 98 | ||
More Information
Set Membership: |
|

LOC101106976
Genomic Map