Assay Details
| Gene Symbol: LOC101113819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961987.1 | XP_027817788.1 | 27 - 28 | 4606 | 67 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101113819| Gene Symbol: LOC101113819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961987.1 | XP_027817788.1 | 27 - 28 | 4606 | 67 | |