Assay Details
| Gene Symbol: LOC101107513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966143.1 | XP_027821944.1 | 1 - 2 | 164 | 82 | |
| XM_027966144.1 | XP_027821945.1 | 1 - 2 | 164 | 82 | ||
More Information
Set Membership: |
|

LOC101107513
Genomic Map