Assay Details
| Gene Symbol: LOC101107513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966143.1 | XP_027821944.1 | 5 - 6 | 413 | 182 | |
| XM_027966144.1 | XP_027821945.1 | 3 - 4 | 314 | 83 | ||
More Information
Set Membership: |
|

LOC101107513
Genomic Map