Assay Details
| Gene Symbol: KANK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969743.1 | XP_027825544.1 | 1 - 2 | 1553 | 65 | |
| XM_027969744.1 | XP_027825545.1 | 1 - 2 | 1553 | 65 | ||
| XM_027969745.1 | XP_027825546.1 | 4 - 5 | 1624 | 65 | ||
| XM_027969746.1 | XP_027825547.1 | 3 - 4 | 1878 | 65 | ||
| XM_027969747.1 | XP_027825548.1 | 3 - 4 | 1878 | 65 | ||
More Information
Set Membership: |
|

KANK3
Genomic Map