Assay Details
| Gene Symbol: LOC114118052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978735.1 | XP_027834536.1 | 1 | 265 | 184 | |
| Gene Symbol: PTGES3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967575.1 | XP_027823376.1 | 2 - 3 | 471 | 184 | |
More Information
Set Membership: |
|

LOC114118052
Genomic Map