Assay Details
| Gene Symbol: CTSD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959254.1 | XP_027815055.1 | 8 - 9 | 1180 | 104 | |
| GenBank mRNA | AF164143.1 | - | 7 - 8 | 988 | 104 | |
More Information
Set Membership: |
|

CTSD
Genomic Map