Assay Details
| Gene Symbol: APCDD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977011.1 | XP_027832812.1 | 3 - 4 | 999 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

APCDD1L| Gene Symbol: APCDD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977011.1 | XP_027832812.1 | 3 - 4 | 999 | 85 | |
Set Membership: |
|