Assay Details
| Gene Symbol: LOC101111827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100358.2 | XP_014955844.1 | 4 - 5 | 983 | 74 | |
| XM_027978710.1 | XP_027834511.1 | 5 - 6 | 1554 | 74 | ||
| XM_027978711.1 | XP_027834512.1 | 5 - 6 | 1554 | 74 | ||
More Information
Set Membership: |
|

LOC101111827
Genomic Map