Assay Details
| Gene Symbol: YWHAQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966509.1 | XP_027822310.1 | 3 - 4 | 594 | 120 | |
| XM_027966511.1 | XP_027822312.1 | 3 - 4 | 537 | 120 | ||
More Information
Set Membership: |
|

YWHAQ
Genomic Map