Assay Details
| Gene Symbol: LOC114108673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975165.1 | XP_027830966.1 | 2 - 3 | 215 | 94 | |
| Gene Symbol: LOC101113605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975163.1 | XP_027830964.1 | 1 - 2 | 125 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027975163.1 transcripts.
Set Membership: |
|

LOC114108673
Genomic Map