Assay Details
| Gene Symbol: LOC101113605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975163.1 | XP_027830964.1 | 7 - 8 | 811 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101113605| Gene Symbol: LOC101113605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975163.1 | XP_027830964.1 | 7 - 8 | 811 | 99 | |
Set Membership: |
|