Assay Details
| Gene Symbol: DNAJB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023545.4 | XP_004023594.3 | 1 | 1356 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAJB7| Gene Symbol: DNAJB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023545.4 | XP_004023594.3 | 1 | 1356 | 94 | |
Set Membership: |
|