Assay Details
| Gene Symbol: GALK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010624.4 | XP_004010673.1 | 5 - 6 | 656 | 72 | |
| XM_004010625.4 | XP_004010674.1 | 5 - 6 | 651 | 72 | ||
| XM_012181591.3 | XP_012036981.1 | 5 - 6 | 477 | 72 | ||
| XM_012181593.3 | XP_012036983.1 | 4 - 5 | 566 | 72 | ||
More Information
Set Membership: |
|

GALK2
Genomic Map