Assay Details
| Gene Symbol: MRPL47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003131.3 | XP_004003180.1 | 6 - 7 | 667 | 97 | |
| XM_012098587.3 | XP_011953977.1 | 6 - 7 | 825 | 97 | ||
More Information
Set Membership: |
|

MRPL47
Genomic Map