Assay Details
| Gene Symbol: LOC101106806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012101164.3 | XP_011956554.1 | 7 - 8 | 1028 | 125 | |
| XM_012101169.3 | XP_011956559.1 | 6 - 7 | 990 | 125 | ||
| XM_027958396.1 | XP_027814197.1 | 5 - 6 | 1088 | 125 | ||
| XM_027958397.1 | XP_027814198.1 | 7 - 8 | 1030 | 125 | ||
More Information
Set Membership: |
|

LOC101106806
Genomic Map