Assay Details
| Gene Symbol: LOC101111723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974885.1 | XP_027830686.1 | 18 - 19 | 2231 | 91 | |
| XM_027974886.1 | XP_027830687.1 | 18 - 19 | 2228 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101111723| Gene Symbol: LOC101111723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974885.1 | XP_027830686.1 | 18 - 19 | 2231 | 91 | |
| XM_027974886.1 | XP_027830687.1 | 18 - 19 | 2228 | 91 | ||