Assay Details
| Gene Symbol: LOC101111723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974885.1 | XP_027830686.1 | 24 - 25 | 3313 | 106 | |
| XM_027974886.1 | XP_027830687.1 | 24 - 25 | 3310 | 106 | ||
More Information
Set Membership: |
|

LOC101111723
Genomic Map