Assay Details
| Gene Symbol: LOC101107096 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969775.1 | XP_027825576.1 | 21 - 22 | 2992 | 82 | |
| XM_027969776.1 | XP_027825577.1 | 20 - 21 | 2845 | 82 | ||
| XM_027969777.1 | XP_027825578.1 | 20 - 21 | 2845 | 82 | ||
| XM_027969778.1 | XP_027825579.1 | 20 - 21 | 2845 | 82 | ||
| XM_027969779.1 | XP_027825580.1 | 20 - 21 | 2836 | 82 | ||
More Information
Set Membership: |
|

LOC101107096
Genomic Map