Assay Details
| Gene Symbol: ANKRD37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012105729.3 | XP_011961119.1 | 3 - 4 | 481 | 91 | |
| XM_027962500.1 | XP_027818301.1 | 3 - 4 | 477 | 91 | ||
More Information
Set Membership: |
|

ANKRD37
Genomic Map