Assay Details
| Gene Symbol: IL20RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011370.4 | XP_004011419.2 | 5 - 6 | 709 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL20RA| Gene Symbol: IL20RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011370.4 | XP_004011419.2 | 5 - 6 | 709 | 80 | |
Set Membership: |
|