Assay Details
| Gene Symbol: EFNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975978.1 | XP_027831779.1 | 3 - 4 | 573 | 87 | |
| XM_027975985.1 | XP_027831786.1 | 3 - 4 | 572 | 87 | ||
More Information
Set Membership: |
|

EFNA4
Genomic Map