Assay Details
| Gene Symbol: RSPO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014778.3 | XP_004014827.2 | 3 - 4 | 408 | 90 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RSPO4| Gene Symbol: RSPO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014778.3 | XP_004014827.2 | 3 - 4 | 408 | 90 | |