Assay Details
| Gene Symbol: PPP4R3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017947.4 | XP_004017996.1 | 5 - 6 | 995 | 74 | |
| XM_012098936.3 | XP_011954326.1 | 5 - 6 | 995 | 74 | ||
More Information
Set Membership: |
|

PPP4R3A
Genomic Map