Assay Details
| Gene Symbol: VPS33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012098474.2 | XP_011953864.2 | 8 - 9 | 944 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VPS33B| Gene Symbol: VPS33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012098474.2 | XP_011953864.2 | 8 - 9 | 944 | 68 | |
Set Membership: |
|