Assay Details
| Gene Symbol: CDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974103.1 | XP_027829904.1 | 1 - 2 | 787 | 92 | |
| XM_027974104.1 | XP_027829905.1 | 1 - 2 | 293 | 92 | ||
More Information
Set Membership: |
|

CDK3
Genomic Map