Assay Details
| Gene Symbol: UNC93B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959506.1 | XP_027815307.1 | 10 - 11 | 1524 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UNC93B1| Gene Symbol: UNC93B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959506.1 | XP_027815307.1 | 10 - 11 | 1524 | 84 | |
Set Membership: |
|