Assay Details
| Gene Symbol: NAAA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012180181.3 | XP_012035571.3 | 6 - 7 | 942 | 66 | |
| XM_027971092.1 | XP_027826893.1 | 6 - 7 | 942 | 66 | ||
More Information
Set Membership: |
|

NAAA
Genomic Map