Assay Details
| Gene Symbol: ARHGEF38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009653.4 | XP_004009702.2 | 10 - 11 | 1668 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARHGEF38| Gene Symbol: ARHGEF38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009653.4 | XP_004009702.2 | 10 - 11 | 1668 | 89 | |
Set Membership: |
|