Assay Details
| Gene Symbol: CDH19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020602.4 | XP_004020651.2 | 2 - 3 | 524 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDH19| Gene Symbol: CDH19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020602.4 | XP_004020651.2 | 2 - 3 | 524 | 90 | |
Set Membership: |
|