Assay Details
| Gene Symbol: CLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012096452.3 | XP_011951842.2 | 2 - 3 | 674 | 109 | |
| XM_027979795.1 | XP_027835596.1 | 2 - 3 | 750 | 109 | ||
| XM_027979796.1 | XP_027835597.1 | 2 - 3 | 1120 | 109 | ||
| XM_027979797.1 | XP_027835598.1 | 3 - 4 | 847 | 109 | ||
| XM_027979798.1 | XP_027835599.1 | 2 - 3 | 822 | 109 | ||
| XM_027979799.1 | XP_027835600.1 | 2 - 3 | 804 | 109 | ||
More Information
Set Membership: |
|

CLP1
Genomic Map