Assay Details
| Gene Symbol: LOC101113671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966011.1 | XP_027821812.1 | 7 - 8 | 795 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101113671| Gene Symbol: LOC101113671 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966011.1 | XP_027821812.1 | 7 - 8 | 795 | 102 | |
Set Membership: |
|